ContactUsLinkComponent
Specifications:- Capillary Column Phase: DB-1ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Form at: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Low Polarity. With Sma 1 * 1 items
Catalog # HEWL122-0132UI-INT
Supplier:  Agilent
undefined
Specifications:- Capillary Column Phase: DB-1ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Form at: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Low Polarity. With Sma 1 * 1 items
Catalog # HEWL122-0132UI-INT
Supplier:  Agilent
Frequently Bought Together