ContactUsLinkComponent
Specifications:- Capillary Column Phase: DB-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.36 um. GC Column Form at: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.18 mm. Length: 20 m. Polarity: Low Polarity. With Sma 1 * 1 items
Catalog # HEWL121-5523UI-INT
Supplier:  Agilent
Specifications:- Capillary Column Phase: DB-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.36 um. GC Column Form at: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.18 mm. Length: 20 m. Polarity: Low Polarity. With Sma 1 * 1 items
Catalog # HEWL121-5523UI-INT
Supplier:  Agilent
Frequently Bought Together