ContactUsLinkComponent
Specifications:- Capillary Column Phase: DB-5ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 items
Catalog # HEWL122-5562-KEY
Supplier:  Agilent
Specifications:- Capillary Column Phase: DB-5ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 items
Catalog # HEWL122-5562-KEY
Supplier:  Agilent
Frequently Bought Together