ContactUsLinkComponent
InfinityLab Poroshell 120 EC-CN, 2.1 x 100 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 3.5 %. E ndcapped: Double. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 items
Catalog # HEWL695775-905T
Supplier:  Agilent
InfinityLab Poroshell 120 EC-CN, 2.1 x 100 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 3.5 %. E ndcapped: Double. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 items
Catalog # HEWL695775-905T
Supplier:  Agilent
Frequently Bought Together