ContactUsLinkComponent
Specifications:- Carbon Load: 10 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: EC-C18. LC Platform: Low Di 1 * 1 items
Catalog # HEWL695775-302
Supplier:  Agilent
Specifications:- Carbon Load: 10 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: EC-C18. LC Platform: Low Di 1 * 1 items
Catalog # HEWL695775-302
Supplier:  Agilent
Frequently Bought Together