ContactUsLinkComponent
InfinityLab Poroshell 120 Phenyl-Hexyl, 2.1, 150 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 items
Catalog # HEWL693775-912T
Supplier:  Agilent
InfinityLab Poroshell 120 Phenyl-Hexyl, 2.1, 150 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 items
Catalog # HEWL693775-912T
Supplier:  Agilent
Frequently Bought Together