ContactUsLinkComponent
InfinityLab Poroshell 120 HILIC, 2.1 x 150 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 0 %. End capped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 items
  HEWL693775-901T
 :  Agilent
InfinityLab Poroshell 120 HILIC, 2.1 x 150 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 0 %. End capped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 items
  HEWL693775-901T
 :  Agilent
Frequently Bought Together