ContactUsLinkComponent
InfinityLab Poroshell HPH-C8, 2.1, 250 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Colum n: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Max 1 * 1 items
Catalog # HEWL690770-706T
Supplier:  Agilent
InfinityLab Poroshell HPH-C8, 2.1, 250 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Colum n: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Max 1 * 1 items
Catalog # HEWL690770-706T
Supplier:  Agilent
Frequently Bought Together