ContactUsLinkComponent
InfinityLab Poroshell 120 PFP, 2.1, 250 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5.1 %. Endcap ped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshe 1 * 1 items
Catalog # HEWL650750-408T
Supplier:  Agilent
InfinityLab Poroshell 120 PFP, 2.1, 250 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5.1 %. Endcap ped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshe 1 * 1 items
Catalog # HEWL650750-408T
Supplier:  Agilent
Frequently Bought Together