ContactUsLinkComponent
ZORBAX RRHD Eclipse PAH, 95A, 3.0 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specifications:- Carbon Load: 14 %. Endcap ped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: ZORBAX. LC Column Max 1 * 3 items
Catalog # HEWL823750-933
Supplier:  Agilent
ZORBAX RRHD Eclipse PAH, 95A, 3.0 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specifications:- Carbon Load: 14 %. Endcap ped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: ZORBAX. LC Column Max 1 * 3 items
Catalog # HEWL823750-933
Supplier:  Agilent
Frequently Bought Together