ContactUsLinkComponent
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 inf inity II Bio LC System. LC Use With Instrument: G7131A. Mixer Material: MP35N. Volume: 35 uL. 1 * 1 items
Catalog # HEWLG7131-68035
Supplier:  Agilent
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 inf inity II Bio LC System. LC Use With Instrument: G7131A. Mixer Material: MP35N. Volume: 35 uL. 1 * 1 items
Catalog # HEWLG7131-68035
Supplier:  Agilent
Frequently Bought Together