SPME Arrow, selection set 2. Includes five different SPME Arrows (20 mm phase length): PDMS (1.10 mm, 100 um, 5191-5862) Polyacryla te (1.10 mm, 100 um, 5191-5858) Carbon WR/PDMS (1.10 mm, 120 um, 5191-5859) DVB/PDMS (1.10 mm, 120 um, 5191-5860) 1 * 1 items
Catalog # HEWL5191-5869
Supplier: Agilent
Frequently Bought Together