ContactUsLinkComponent
Specifications:- Compatible Column ID: 0.32 mm. Compatible Column OuterDiameter (OD): 0.45 mm. Ferrule Material: Stainless steel. Fe rrule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.5 mm. Manufacturer Compatibility: Agilent. 1 * 10 items
Catalog # HEWLG2855-28502
Supplier:  Agilent
Specifications:- Compatible Column ID: 0.32 mm. Compatible Column OuterDiameter (OD): 0.45 mm. Ferrule Material: Stainless steel. Fe rrule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.5 mm. Manufacturer Compatibility: Agilent. 1 * 10 items
Catalog # HEWLG2855-28502
Supplier:  Agilent
Frequently Bought Together