Static dissipative facemasks
L/XL, Blue
Catalog # DESC97553
Supplier: DESCO
Jump to:
Product Details & Documents
These facemasks constructed of a lightweight dissipative material which is made from texturised polyester and a minimum of 9% carbon nylon monofiliment.
- Can be washed multiple times in a mild detergent
- Elastic ear loops
- Adjustable nose wire helps ensure mask fits securely
- Acceptable for use in ESD protected areas
- Provides continuous and consistent charge dissipation
Elastic bands range from small-medium (69,85 mm) to large-extra large (82,55 mm). The mask area is the same size for all items.
Specifications
- Colour:Blue
- Seq:04
- Size:L/XL
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.