Static dissipative facemasks
S/M, Grey
N° de catalogue DESC97552
Fournisseur: DESCO
Jump to:
Product Details & Documents
These facemasks constructed of a lightweight dissipative material which is made from texturised polyester and a minimum of 9% carbon nylon monofiliment.
- Can be washed multiple times in a mild detergent
- Elastic ear loops
- Adjustable nose wire helps ensure mask fits securely
- Acceptable for use in ESD protected areas
- Provides continuous and consistent charge dissipation
Elastic bands range from small-medium (69,85 mm) to large-extra large (82,55 mm). The mask area is the same size for all items.
Specifications
- Colour:Grey
- Seq:03
- Size:S/M