Orders LinkContactUsLinkComponent
Embedding rings
Embedding rings
Yellow
Catalog # KART0291906
Supplier:   KARTELL
Embedding rings
Catalog # KART0291906
Supplier:  KARTELL
Embedding rings
Embedding rings
Yellow
Price
370,00 €
Log in for availability
1 * 2.000 ST
Product Details & Documents
Embedding rings made from ABS.
  • Dimensions: 41×28×11 mm
Available in the colours white, blue, yellow, green and pink.
Specifications
  • Colour:
    Yellow