Micro Test Tubes, Graduated
Micro test tube, graduated, 1,5 ml, Green, PP
KART0027912
: KARTELL
Ideal for samples analysis.
- Made from PP
- Flat cap with writing area
- Available in blue, yellow, pink and green
Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11000.
Certifications: The raw materials with the relevant requirements of Food-contact EU; Regulation 1935/2004 EC, Regulation 2023/2006/EC, FDA and REACH/ROHS regulations.
- Capacity:1,5 ml
- Colour:Green
- Description:Micro test tube, graduated
- Material:PP
- Seq:03