Micro Test Tubes, Graduated
Micro test tube, graduated, 1,5 ml, Yellow, PP
Catalog # KART0027906
Supplier: KARTELL
Jump to:
Product Details & Documents
Ideal for samples analysis.
- Made from PP
- Flat cap with writing area
- Available in blue, yellow, pink and green
Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11000.
Certifications: The raw materials with the relevant requirements of Food-contact EU; Regulation 1935/2004 EC, Regulation 2023/2006/EC, FDA and REACH/ROHS regulations.
Specifications
- Capacity:1,5 ml
- Colour:Yellow
- Description:Micro test tube, graduated
- Material:PP
- Seq:02
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.