Orders LinkContactUsLinkComponent
VWR®, Cryo coder cap inserts
VWR®, Cryo coder cap inserts
Cap insert, Pink
Catalog # VWRI210-000042
Supplier:   VWR Collection
VWR®, Cryo coder cap inserts
Catalog # VWRI210-000042
Supplier:  VWR Collection
VWR®, Cryo coder cap inserts
VWR®, Cryo coder cap inserts
Cap insert, Pink
Price
23,80 €
Log in for availability
1 * 500 ST
Product Details & Documents
Product can be used as coding/identification of samples types stored in it.
Cryocoder/cap inserts are available in multiple colour options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).
Specifications
  • Colour:
    Pink
  • Description:
    Cap insert
  • Seq:
    07