Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Ointment jars, with thread

Ointment jar, clear glass, 30 ml, 36 mm, GL 46
  216-2972
Ointment jars, with thread
  216-2972
Ointment jars, with thread
Ointment jars, with thread

Ointment jar, clear glass, 30 ml, 36 mm, GL 46

 
146,00 €
   
1 * 90 ST
 

Made from clear or amber soda-lime glass.

  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • With round thread

Ideal for sampling, storage and transportation of powders and pastes. Amber jars to protect light-sensitive materials.

 : Screw caps have to be ordered separately.

 
  • Thread:
    GL 46
  • Capacity:
    30 ml
  • Description:
    Ointment jar, clear glass
  • Height:
    36 mm
  • Seq:
    01
Accessories

 

  •  
    Description
     
     
  • RIXI10-23-41
    White polypropylene insert lid for 30 ml jars with thread 46
       
    1 * 10 ST - 1,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.