AMMONIUM MOLBYDATE TETRAHYDRATE CRYSTAL 1 * 500 g
Catalog # SPCMA1210-500GM
Supplier: Spectrum Chemical
AMMONIUM MOLBYDATE TETRAHYDRATE CRYSTAL 1 * 500 g
Catalog # SPCMA1210-500GM
Supplier: Spectrum Chemical
Frequently Bought Together
Sodium dodecyl sulphate (SDS) ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
EDTA disodium salt dihydrate BP, Ph. Eur., ACS, JP, USP
Supplier: VWR Chemicals
Ammonium sulphate, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium hexacyanoferrate(III) ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium chromate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium nitrate 99+% for analysis
Supplier: Thermo Fisher Scientific
Lithium tetrafluoroborate, anhydrous 98%
Supplier: Apollo Scientific
Lithium Carbonate (99.999%-Li) Puratrem 1 * 100 g
Supplier: Strem Chemicals, Inc.