(2R,3R,4R,5R)-5-(2-AMINO-6-CHLORO-9H-PUR 1 * 10 g
Catalog # MSPPAR000ZTR_10
Supplier: MARKET SOURCE PART PROCESS
(2R,3R,4R,5R)-5-(2-AMINO-6-CHLORO-9H-PUR 1 * 10 g
Catalog # MSPPAR000ZTR_10
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetonitrile 99.9% ACS, gradient grade, meets analytical specification of Ph.Eur. for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
Diethyl ether
Supplier: Roth Carl
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Acetone, BAKER, J.T.Baker®
Supplier: Avantor
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: Honeywell Chemicals