SODIUIM HYDROXIDE 50% SOLUTION 1 * 500 mL
SPCMS-224-500ML
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
: VWR Chemicals
Kjeldahl tablets
: Merck
FORMALDEHYDE CONCENTRATE 24% 1 * 10 L
: KLINIPATH
BUFFER pH 2 ACC. TO NF T 90-431 1 * 1 L
: PROLABO CMP