Order Entry
Belgium
Orders LinkContactUsLinkComponent
Recycling solution for plastic packaging, Zero Waste Box™

Recycling solution for plastic packaging, Zero Waste Box™

Disposal box for plastic packaging, small, 28 L, ≤7 kg, 250×250×460 mm
Catalog # 129-2557
   Product Transparency and Disclosure|Waste Reducing
Supplier:   TERRACYCLE
Recycling solution for plastic packaging, Zero Waste Box™
Catalog # 129-2557
Supplier:  TERRACYCLE
   Product Transparency and Disclosure|Waste Reducing
Recycling solution for plastic packaging, Zero Waste Box™
Recycling solution for plastic packaging, Zero Waste Box™

Disposal box for plastic packaging, small, 28 L, ≤7 kg, 250×250×460 mm

Price
111,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

A Zero WasteBox™ is a complete and convenient solution which includes the storage, shipping and recycling of a particular type of waste that isn’t currently recycled through local councils or traditional recycling facilities.

  • Reduce the amount of waste that you send to landfill or incinerate
  • Recycle the non-recyclable and reduce your carbon footprint
  • Improving your organisation's sustainability can boost your corporate image and improve employee engagement and satifaction

Use this Zero WasteBox™ to recycle any flexible plastic packaging, carrier bags and shipping material including bubble wrap, polystyrene, airbags, shrink wrap, cushioning pallet liners and covers. Any hard / rigid plastic packaging and plastic containers. Use this box to recycle VWR® PureStep EnviroTack™ mat layers, including top and bottom release liners.

How it works;
Order - Select a Zero WasteBox™ based on what and how much you would like to recycle.
Collect - Fill your box with the accepted waste and seal the box with adhesive tape
Ship - Using the pre-paid UPS shipping label affixed to the back of your box, send your filled box to TerraCycle® for recycling. UPS requires a minimum of 24 hours’ notice and does pick ups Monday to Friday between 9am and 5pm. Pick-ups can be arranged online or by telephone. All shipments are carbon-neutral, meaning the carbon emissions released during transport will be offset.
Feel free to contact UPS on 078 250 877 or ups.com/be/fr
Reorder - Continue to recycle and reorder a new Zero WasteBox™ . Your collected waste will find a second life as recycled materials, reducing the need to extract virgin materials and depleting resources.

Cardboard boxes are made from 84% recycled material. Box container and liner are 100% recyclable.

Certifications: TerraCycle® will not landfill or incinerate any of the waste received.

Caution: Do not send in food content and liquid waste. Plant-based or biodegradable plastics (e.g. PLA). Any non-plastic waste: paper, metals, cardboard, glass. Plastic cards. Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people. Batteries, pressurised canisters, broken glass and medical waste.

Specifications
  • Capacity:
    28 L
  • Description:
    Disposal box for plastic packaging, small
  • Seq:
    01
  • W×D×H:
    250×250×460 mm
  • Load capacity:
    ≤7 kg
  • Cat. No.:
    129-2557
  • Environmental certification:
    FSC (Certificate code FSC: BV-COC-150648), Bureau Veritas, Third party verified LCA
  • Environmentally Preferable:
    Product Transparency and Disclosure|Waste Reducing
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.