Orders LinkContactUsLinkComponent
Cleanroom boot covers, CleanMax®
Cleanroom boot covers, CleanMax®
Clean manufactured boot cover, White, S/M
Catalog # 114-9547
Cleanroom boot covers, CleanMax®
Catalog # 114-9547
Cleanroom boot covers, CleanMax®
Cleanroom boot covers, CleanMax®
Clean manufactured boot cover, White, S/M
Price
453,00 €
Log in for availability
1 * 50 PAIRE
Product Details & Documents
Clean manufactured disposable boot covers are individually packaged in pairs and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • 19" high
  • Non-skid PVC sole
  • Individually bagged in pairs
  • Latex-free
  • Tunnelled elastic top
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparel provides chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all controlled environments.
Specifications
  • Shoe size:
    S/M
  • Colour:
    White
  • Description:
    Clean manufactured boot cover
  • Seq:
    01
  • Cat. No.:
    114-9547
  • Garment application:
    Cleanroom