Orders LinkContactUsLinkComponent
Containers with Sloping Base
Containers with Sloping Base
Hygienic valve kit with silicone seals, —
Catalog # SAMPA136-01S
Supplier:   Sampling Systems
Containers with Sloping Base
Catalog # SAMPA136-01S
Supplier:  Sampling Systems
Containers with Sloping Base
Containers with Sloping Base
Hygienic valve kit with silicone seals, —
Price
425,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These drums which incorporate a sloping base allow the container to drain completely.
  • Made from 316L stainless steel
  • Includes lid, clamp and silicone seal
  • Other seals available
  • Ferrule for easy emptying
  • Handles enable easy transportation
The Sampling Systems range of containers with sloping base ensures that material can be completely drained from within and include a ferrule at the base for easy emptying. They have a mirror polish finish for easy cleaning.
Specifications
  • Capacity:
  • Description:
    Hygienic valve kit with silicone seals
  • Seq:
    06