Increase confidence in your cell thawing workflow by ensuring sample sterility and consistent thawing performance by using the ThawS TAR reg CFT2 Automated Thawing System. With a standardized thawing process that replaces manual, water bath-based th 1 * 20 items
Catalog # STMC100-0643
Supplier: STEMCELL Technologies
Frequently Bought Together