Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Human/Mouse Recombinant BMP-2, ACF

10 µG
  STMC78135
Marketsource
Human/Mouse Recombinant BMP-2, ACF
  STMC78135
Marketsource
 :  
Human/Mouse Recombinant BMP-2, ACF
Human/Mouse Recombinant BMP-2, ACF

10 µG

 :  
 
214,00 €
   
1 * 1 EA
 

Bone morphogenetic protein 2 (BMP-2) is a member of the transforming growth factor beta (TGF-β) superfamily. BMP-2 is a disulfide-linked homodimer, acts as a ligand for complexes of type I and II BMP receptors, and primarily activates SMAD1/5/8 signaling (Nohe et al.). BMP-2 is a potent differentiation factor and directs human pluripotent stem cells (hPSCs) towards various cell types including extra-embryonic endoderm, mesenchymal cells, and chondrocytes (Pera et al.). Although BMP-2 expression is low in healthy cartilage, its expression is upregulated at the site of cartilage damage (Blaney Davidson et al.). BMP-2 induces bone and cartilage formation in vitro and is able to induce chondrogenesis in human mesenchymal stem cells (Schmitt et al.). This product is animal component-free.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

 

 

  • Pk:
    10 µG
  • Cat. No.:
    STMC78135
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    BMP-2, ACF
  • Purity:
    ≥95%
  • Species:
    Human|Mouse