ContactUsLinkComponent
40X WATER IMMERSION SYSTEM INCLUDES 40X APO LWD 1.15 NA WATER IMMERSION OBJECTIVE AND DISPENSER MODULE 1 * 1 items
Catalog # MLDV5076809
Supplier:  Molecular Devices
40X WATER IMMERSION SYSTEM INCLUDES 40X APO LWD 1.15 NA WATER IMMERSION OBJECTIVE AND DISPENSER MODULE 1 * 1 items
Catalog # MLDV5076809
Supplier:  Molecular Devices
Frequently Bought Together