Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Septa ND11

VWR®, Septa ND11

Grey PTFE/cream butyl/grey PTFE, 55° shore A, 1,3 mm
Catalog # 548-0843A
Supplier:   VWR Collection
VWR®, Septa ND11
Catalog # 548-0843A
Supplier:  VWR Collection
VWR®, Septa ND11
VWR®, Septa ND11

Grey PTFE/cream butyl/grey PTFE, 55° shore A, 1,3 mm

Price
163,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.

  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available

Septa choice depends on the application.

Specifications
  • Hardness:
    55° shore A
  • Septum:
    Grey PTFE/cream butyl/grey PTFE
  • Seq:
    08
  • Thickness:
    1,3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.