ContactUsLinkComponent
Masterflex® I/P® Easy-Load® Replacement Rotor and Bearing Assembly for 77602-12 Pump Head
Masterflex® I/P® Easy-Load® Replacement Rotor and Bearing Assembly for 77602-12 Pump Head
Catalog # MFLX77602-38
undefined
Masterflex® I/P® Easy-Load® Replacement Rotor and Bearing Assembly for 77602-12 Pump Head
Catalog # MFLX77602-38
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
  • Suitable for use with I/P® Easy-Load® pump heads
Replacement parts to keep your pump head operating even longer.
  • Pump series:
    I/P
  • Description:
    I/P® Easy-Load® Replacement Rotor and Bearing Assembly for 77602-12 Pump Head
  • Environmentally Preferable:
  • Warranty:
    3 Months
  • Pk:
    1
  • Housing material:
    PPS
  • Rotor material:
    Stainless Steel
Frequently Bought Together