(1R,2R)-1,2- BIS((DIPHENYLPHOSPHINO)METH 1 * 250 mg
Catalog # AMBDA458339-250MG
Supplier: AMBEED
Frequently Bought Together
1-Octanol, EMPLURA®, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥98%, TECHNICAL
Supplier: VWR Chemicals
1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dodecylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, Bioreagent
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.9%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
4-(Dimethylamino)benzaldehyde, Supelco®
Supplier: Merck
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals