ContactUsLinkComponent
Human BCHE, His Tag (BHE-H52H3) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Leu 602 (Accession # P06276-1). 1 * 500 µG
  ACRBBHE-H52H3500UG
Human BCHE, His Tag (BHE-H52H3) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Leu 602 (Accession # P06276-1). 1 * 500 µG
  ACRBBHE-H52H3500UG
Frequently Bought Together