ContactUsLinkComponent
GMP Human IL-7 (GMP-L07H24) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession # P13232-1). 1 * 50 µG
Catalog # ACRBGMP-L07H2450UG
Supplier:  ACROBIOSYSTEMS
GMP Human IL-7 (GMP-L07H24) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession # P13232-1). 1 * 50 µG
Catalog # ACRBGMP-L07H2450UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together