ContactUsLinkComponent
Varicella zoster virus (strain Oka vaccine) Envelope Glycoprotein E (gE), His Tag (GLE-V52H3) is expressed from human 293 cells (HEK 293). It contains AA Ser 31 - Ala 546 (Accession # Q9J3M8-1). 1 * 100 µG
Catalog # ACRBGLE-V52H3100UG
Supplier:  ACRO Biosystems
Varicella zoster virus (strain Oka vaccine) Envelope Glycoprotein E (gE), His Tag (GLE-V52H3) is expressed from human 293 cells (HEK 293). It contains AA Ser 31 - Ala 546 (Accession # Q9J3M8-1). 1 * 100 µG
Catalog # ACRBGLE-V52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together