BIOTINYLATED HUMAN INTEGRIN ALPHA V BETA 3 (ITGAV&ITGB3) HETERODIMER PROTEIN, HIS,AVITAG &TAG FREE 1 * 200 µG
Catalog # ACRBIT3-H82W9200UG
Supplier: ACRO Biosystems
BIOTINYLATED HUMAN INTEGRIN ALPHA V BETA 3 (ITGAV&ITGB3) HETERODIMER PROTEIN, HIS,AVITAG &TAG FREE 1 * 200 µG
Catalog # ACRBIT3-H82W9200UG
Supplier: ACRO Biosystems
Frequently Bought Together
Laboratory trays
Supplier: KARTELL
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Deep well storage plates, 96-well, 1,2 ml
Supplier: THERMO MATRIX TECHNOLOGIES
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cryo.S™ tubes, 1 and 2 ml
Supplier: Greiner Bio-One
VWR®, Sample Containers, with Attached Lids
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Corning® CoolCell® containers
Supplier: Corning
VWR®, Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Supplier: EPPENDORF
Human recombinant integrin alpha V beta 6 (from HEK293)
Supplier: ACROBIOSYSTEMS
Electroporation cuvettes
Supplier: Thermo Fisher Scientific
Accessories and consumables for KingFisher™ Flex
Supplier: Thermo Fisher Scientific
Measuring scoops
Supplier: VITLAB