ContactUsLinkComponent
Human Laminin 221 E8, Fc Tag (LA2-H5269) is expressed from human 293 cells (HEK293). It contains AA Leu 1900 - Ala 2722 (Laminin alp ha2E8) & Leu 1573 - Gln 1798 (Laminin beta2E8) & Asn1364 - Pro1609 (Laminin gamma1E8) (Accession # P24043-1 (Laminin 1 * 1 mg
  ACRBLA2-H5269-1MG
Human Laminin 221 E8, Fc Tag (LA2-H5269) is expressed from human 293 cells (HEK293). It contains AA Leu 1900 - Ala 2722 (Laminin alp ha2E8) & Leu 1573 - Gln 1798 (Laminin beta2E8) & Asn1364 - Pro1609 (Laminin gamma1E8) (Accession # P24043-1 (Laminin 1 * 1 mg
  ACRBLA2-H5269-1MG
Frequently Bought Together