ContactUsLinkComponent
Canine FCGRT&B2M Heterodimer Protein, His Tag&Tag Free (FCM-C52W8) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Ser 298 (FCGRT) & Val 28 - Asn 125 (B2M) (Accession # XP_038512242.1 (FCGRT) & NP_001271408.1 (B2M)). 1 * 1 mg
Catalog # ACRBFCM-C52W8-1MG
Supplier:  ACRO Biosystems
Canine FCGRT&B2M Heterodimer Protein, His Tag&Tag Free (FCM-C52W8) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Ser 298 (FCGRT) & Val 28 - Asn 125 (B2M) (Accession # XP_038512242.1 (FCGRT) & NP_001271408.1 (B2M)). 1 * 1 mg
Catalog # ACRBFCM-C52W8-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together