ContactUsLinkComponent
Masterflex® Fitting, PVDF, Straight, Threaded Reducer, 1/8" NPT(M) x 3/8" NPT(M); 10/PK
Masterflex® Fitting, PVDF, Straight, Threaded Reducer, 1/8" NPT(M) x 3/8" NPT(M); 10/PK
Catalog # MFLX40615-32
undefined
Masterflex® Fitting, PVDF, Straight, Threaded Reducer, 1/8" NPT(M) x 3/8" NPT(M); 10/PK
Catalog # MFLX40615-32
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ⅛"
  • Thread size NPT(M) (port 2):
    ⅜"
  • Description:
    Fitting, PVDF, Straight, Threaded Reducer, ⅛" NPT(M) x ⅜" NPT(M); 10/PK
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Pk:
    10
  • Fitting material:
    PVDF
Frequently Bought Together