Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Reactor systems, EasySyn
Reactor systems, EasySyn
EasySyn 2000 starter reactor system, 2 L
Catalog # IKAA20113438
Supplier:   IKA
Reactor systems, EasySyn
Catalog # IKAA20113438
Supplier:  IKA
Reactor systems, EasySyn
Reactor systems, EasySyn
EasySyn 2000 starter reactor system, 2 L
Price
11 860,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
EasySyn is the reactor system for organic or aqueous syntheses, and the only reactor system for which you can get all the complementary components from a single source: Overhead stirrers, circulators, vacuum pumps and controllers, sensors and automation software. All from IKA and all perfectly matched to each other. With its double-walled reactor vessels from 500 to 5000 ml, EasySyn is a medium-sized and very flexible upscaling system. It ensures safe and reproducible chemical reactions.
  • Ideal for organic or aqueous syntheses
The EasySyn reactor systems are characterised by a wide range of applications such as reflux condensations, reactions under vacuum, crystallisations, (pH-controlled) precipitation reactions, nanoparticle or catalyst syntheses as well as the transfer from laboratory to process scale.
In many applications, controllable and reproducible reaction conditions are essential to achieve reliable and comparable results. With EasySyn, these conditions are guaranteed by the use of high-quality materials in combination with technically sophisticated accessories. Our reactor systems are for example suitable for research and development, the chemical industry or materials research.
Ergonomic and safe - They are unique thanks to the ergonomically designed stand. It allows quick and easy exchange of the reactor vessels, simple alignment of the stirrer and adjustment of the working height to the user's body size.
Due to the use of borosilicate glass 3.3 and PTFE-coated stirring tools, EasySyn has a high chemical resistance and can also be used for syntheses with aggressive media such as acids or bases. The high quality of the materials used allows operation at temperatures from −50 to 200 °C and up to a vacuum of 3 mbar.
One system, many possibilities - In total, two packages are offered, each with different sizes and variations: EasySyn Starter and EasySyn Advanced. They each comprise a complete reactor system for use in the laboratory, consisting of a stand system, double-walled reactor vessel, reactor lid as well as a propeller stirrer and a suitable overhead stirrer.
The main difference between Starter and Advanced is the overhead stirrer used. EasySyn Starter includes the EUROSTAR 60 digital, the Advanced Packages the EUROSTAR 100 control, which enables additional functions, such as external control via labworldsoft® 6 (RS232 or USB interface), operation via removable remote control and programming, for example, of a timer. In addition, the Advanced systems include the LT 5.40 draining set, which enables the tempering fluid of the double jacket to be drained easily and cleanly by using special valves, thus saving valuable time when changing the reactor vessels. The Advanced System is therefore particularly suitable for applications in which the reactor vessel has to be changed or cleaned regularly.
Areas of application: Organic and aqueous syntheses; Reactions under vacuum; Crystallisations; (pH-controlled) precipitation reactions; Nanoparticle syntheses; Catalyst syntheses. Matching to this you get all the complementary components from a single source: overhead stirrers, circulators, vacuum pumps and controllers, sensors and automation software.
Specifications
  • Désignation:
    EasySyn 2000 starter reactor system, 2 L
  • Seq:
    05
  • Cat. No.:
    IKAA20113438
  • L×P×H:
    500×580×950 mm
  • Volume utile:
    120 ml (min.)
  • Vide:
    3 mbar (attainable)
  • Modèle:
    EasySyn 2000
  • Plage de température de fonctionnement:
    5...40 °C (permissible ambient temperature)
Accessories
Product Information
  • Catalog #
    Désignation
    Pour
    Availability
    Price
  • IKAA20103316
    SY stand
    Log in for availability
    1 * 1 ST - 4 530,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • IKAA3980000
    Vacuum pump, MVP 10 basic
    Log in for availability
    1 * 1 ST - 2 380,00 €
Product Information
  • Catalog #
    Type
    Plage de température
    Viscosité
    Availability
    Price
  • IKAA20003521
    Heating fluid
    +20...+200 °C
    50 mm²/s
    Log in for availability
    1 * 10 kg - 761,00 €
Product Information
  • Catalog #
    Désignation
    Pour
    Availability
    Price
  • IKAA20116965
    Reactor lid for the EasySyn reactor system, openings: 4×NS 29/32, 1×NS 14/23, 2×GL 18
    Suitable for reactor vessels with volumes of 500 ml (SY 500 and SY 500 D) and 1000 ml (SY 1000 and SY 1000 D)
    Log in for availability
    1 * 1 ST - 1 090,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • IKAA20016236
    Membrane vacuum pump, VACSTAR digital
    Log in for availability
    1 * 1 ST - 3 420,00 €
Product Information
  • Catalog #
    Type
    Précision de temp.
    Puissance de chauffe
    Availability
    Price
  • IKAA4165000
    CBC 5 basic
    ±0,05 K
    2,5 kW
    Log in for availability
    1 * 1 ST - 6 850,00 €
Product Information
  • Catalog #
    Type
    Plage de température
    Viscosité
    Availability
    Price
  • IKAA20003520
    Heating fluid
    +20...+200 °C
    50 mm²/s
    Log in for availability
    1 * 10 kg - 681,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • 441-0310
    Agitateur EUROSTAR 100 control, prise EU
    Log in for availability
    1 * 1 ST - 3 420,00 €
Product Information
  • Catalog #
    Type
    Plage de température
    Viscosité
    Availability
    Price
  • IKAA20003518
    Heating fluid
    +20...+200 °C
    50 mm²/s
    Log in for availability
    1 * 9 kg - 714,00 €
Product Information
  • Catalog #
    Type
    Précision de temp.
    Puissance de chauffe
    Availability
    Price
  • IKAA4167000
    CBC 5 control
    ±0,05 K
    2,5 kW
    Log in for availability
    1 * 1 ST - 8 170,00 €
Product Information
  • Catalog #
    Désignation
    Pour
    Availability
    Price
  • IKAA10011654
    Hose set includes M 16×1 connection, PUR 90 °C, Ø 12 mm
    Log in for availability
    1 * 1 ST - 205,00 €
Product Information
  • Catalog #
    Désignation
    Pour
    Availability
    Price
  • IKAA4569300
    Tube FKM, 2×1,5 m, M 16×1, −30...+180 °C, Ø 12 mm
    Log in for availability
    1 * 1 ST - 1 070,00 €
Product Information
  • Catalog #
    Désignation
    Pour
    Availability
    Price
  • IKAA20113271
    Temperature control hose, 1,6 m
    Log in for availability
    1 * 1 ST - 1 150,00 €
Product Information
  • Catalog #
    Désignation
    Type
    Précision de temp.
    Availability
    Price
  • IKAA25004524
    HRC 2 temperature control
    HRC 2 control
    0,05±K
    Log in for availability
    1 * 1 ST - 7 170,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • IKAA20117772
    H 71.32 Temperature sensor 320 mm, PTFE coated, Ø 8 mm
    Log in for availability
    1 * 1 ST - 662,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • 441-0326
    Agitateur EUROSTAR 60 digital, prise EU
    Log in for availability
    1 * 1 ST - 1 860,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • 441-0323
    Agitateur EUROSTAR 60 control, prise EU
    Log in for availability
    1 * 1 ST - 2 700,00 €
Product Information
  • Catalog #
    Type
    Précision de temp.
    Puissance de chauffe
    Availability
    Price
  • 441-0408
    HBC 5 control
    ±0,01 K
    2,5 kW
    Log in for availability
    1 * 1 ST - 5 890,00 €
Product Information
  • Catalog #
    Type
    Précision de temp.
    Puissance de chauffe
    Availability
    Price
  • 441-0409
    HBC 5 basic
    ±0,02 K
    2,5 kW
    Log in for availability
    1 * 1 ST - 4 580,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • IKAA20118833
    H 71.45 Temperature sensor 450 mm, PTFE coated, Ø 8 mm
    Log in for availability
    1 * 1 ST - 717,00 €
Product Information
  • Catalog #
    Désignation
    Pour
    Availability
    Price
  • IKAA20113649
    Reactor lid for the EasySyn reactor system, openings: 3×NS 29/32, 1×NS 14/23
    Suitable for reactor vessels with volumes of 500 ml (SY 500 and SY 500 D) and 1000 ml (SY 1000 and SY 1000 D)
    Log in for availability
    1 * 1 ST - 649,00 €
Product Information
  • Catalog #
    Désignation
    Pour
    Availability
    Price
  • IKAA20017366
    Labworldsoft® 6 Pro, with access to the entire device library
    All configurations with any number of devices can be controlled at the same time
    Log in for availability
    1 * 1 ST - 14 820,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • IKAA20121894
    Vanne de vidange inférieure avec sonde de température PT 100 intégrée
    Log in for availability
    1 * 1 ST - 1 220,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.