METHYLENE BLUE 1% IN AQUEOUS SOLUTION 1 * 10 L
Catalog # PRLS89614.410
Supplier: PROLABO CMP
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Test strips, nitrite, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR®, Immersion Oil
Supplier: VWR Collection
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Trypan blue 0,4% in aqueous solution, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Powder pillows, ammonia
Supplier: Hach
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach