1-OCTENE 1 * 100 mg
Catalog # 78340-100MG
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH Standard Solutions, IUPAC, Certified
Supplier: Hach