ContactUsLinkComponent
Masterflex® Quick-Disconnect Body, Acetal, 1/8" Flow, Valved, 1/4" Hose Barb
Masterflex® Quick-Disconnect Body, Acetal, 1/8" Flow, Valved, 1/4" Hose Barb
Catalog # MFLX70000-27
undefined
Masterflex® Quick-Disconnect Body, Acetal, 1/8" Flow, Valved, 1/4" Hose Barb
Catalog # MFLX70000-27
About this item
Disconnect and reconnect your flow path with ease.
  • Audible click lets you know the coupling is secure
  • Many hose barb sizes to choose from
  • Valved design allows you to cut off fluid flow when the fittings are disconnected
  • Material of construction is acetal│Has a 1/4" hose barb
Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and the ability to be gamma sterilized.
  • Barb size/tubing int.Ø (port 1):
    0,25"
  • Connection Type:
    Body
  • Description:
    Masterflex Quick-Disconnect Body, Acetal, ⅛" Flow, Valved, ¼" Hose Barb
  • Seq:
    04
  • Environmentally Preferable:
  • Model:
    70000-27
  • Pk:
    1
  • Fitting material:
    Acetal
  • Quick-disconnect flow size:
    0,13"
Frequently Bought Together