ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Low-Profile Hose Barb Adapter, 11/64" ID; 25/PK
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Low-Profile Hose Barb Adapter, 11/64" ID; 25/PK
Catalog # MFLX45518-13
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Low-Profile Hose Barb Adapter, 11/64" ID; 25/PK
Catalog # MFLX45518-13
CE certificate
About this item
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male Luer Lock to Low-Profile Hose Barb, 11/64" ID
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilisation.
  • Barb size/tubing int.Ø (port 1):
    11/64"
  • Flow pattern:
    Straight
  • Description:
    Luer Adapters, Male Luer Lock to 11/64" ID low profile barb, PP, 25/Pk
  • Seq:
    14
  • Environmentally Preferable:
  • Model:
    45518-13
  • Connection type (port 1):
    Male luer lock
  • Connection type (port 2):
    Hose barb
  • No. of ports:
    2
  • Pk:
    25
  • Fitting material:
    Polypropylene
Frequently Bought Together