ContactUsLinkComponent
Replacement easy clean/decon belt for RPB PX5 PAPR. Allows for easy and efficent decontamination and cleaning of belt when used in a rea's when infection control or cleanliness is of vital importance 1 * 1 items
Catalog # RPBS07-765-DC
Supplier:  RPB SAFETY
Replacement easy clean/decon belt for RPB PX5 PAPR. Allows for easy and efficent decontamination and cleaning of belt when used in a rea's when infection control or cleanliness is of vital importance 1 * 1 items
Catalog # RPBS07-765-DC
Supplier:  RPB SAFETY
Frequently Bought Together