Rubidium Bromide (99%-Rb) 1 * 50 g
Catalog # STRE93-3720-50G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Potassium hexacyanoferrate(II) trihydrate ≥96%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Zinc iodide and starch solution analytical reagent, Supelco®
Supplier: Merck
(±)-α-Methoxyphenylacetic acid Reag. Ph. Eur., Supelco®
Supplier: Merck
Potassium peroxodisulphate, EMSURE® for analysis, Supelco®
Supplier: Merck
Tin(II) chloride dihydrate ≥96%, GPR RECTAPUR® for mirror makers
Supplier: VWR Chemicals
Manganese(II) chloride tetrahydrate 99.0-101.0%, AnalaR NORMAPUR® ACS analytical reagent
Supplier: VWR Chemicals
Ammonium thiocyanate ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Calcium chloride dihydrate 99.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonium hydrogen difluoride ≥98.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium nitrate, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Calcium chloride dihydrate 97.0-103.0% Ph. Eur.
Supplier: VWR Chemicals
Cobalt Naphthenate, In Mineral Spirits ( 1 * 2 kg
Supplier: Strem Chemicals, Inc.
Zinc acetate dihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
di-Sodium L(+)-tartrate dihydrate 99+% ACS
Supplier: Thermo Fisher Scientific
EDTA magnesium dipotassium salt 97%
Supplier: Thermo Fisher Scientific