Titanium Boride, 99% 1 * 100 g
Catalog # STRE93-2242-100G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Calcium chloride dihydrate 99.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Aluminium potassium sulphate dodecahydrate ≥97%, TECHNICAL
Supplier: VWR Chemicals
Sodium azide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium peroxide ≥93%, granules, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium salicylate, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium sulphate, anhydrous 98.5-101.0%, fine powder USP, Ph. Eur.
Supplier: VWR Chemicals
Lithium carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Calcium carbonate ≥99%, GPR RECTAPUR®, precipitated
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Charcoal activated, Supelco®
Supplier: Merck
Sodium carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium chloride 99.0-100.5% ACS
Supplier: Thermo Fisher Scientific
di-Ammonium oxalate monohydrate 99.0-101.0% ACS
Supplier: Thermo Fisher Scientific