Titanium(Iii) Fluoride, Min. 98% 1 * 25 g
Catalog # STRE93-2224-25G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Sodium acetate anhydrous EMPROVE ESSENTIAL ChP,USP,FCC,E 262 1 * 25 kg
Supplier: MERCK PRODUCTION CHEMICALS
Copper ≥99.5%, turnings, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 99%, extra pure
Supplier: Thermo Fisher Scientific
Antimony Tin Oxide/Iridium Het-Woc Core/ 1 * 1 g
Supplier: Strem Chemicals, Inc.
Sodium dihydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
Supplier: VWR Chemicals
Ammonium hydrogen difluoride 95% remainder mainly ammonium fluoride
Supplier: Thermo Fisher Scientific
Silver Telluride (99.99%-Ag/Te) Puratrem 1 * 5 g
Supplier: Strem Chemicals, Inc.
Rhodium Nanoparticles, Pure, (<20Nm) In 1 * 25 mL
Supplier: Strem Chemicals, Inc.
Potassium trimethylsilanolate 90%
Supplier: Apollo Scientific
Sodium chloride, extra pure
Supplier: Thermo Fisher Scientific
Iridium Black (99.9% Ir) 1 * 2 g
Supplier: Strem Chemicals, Inc.
Ammonium peroxodisulphate (APS) ≥98%
Supplier: Thermo Fisher Scientific
Barium Iodide, Anhydrous, Min. 97% 1 * 25 g
Supplier: Strem Chemicals, Inc.
Potassium permanganate ≥99% ACS
Supplier: VWR Chemicals
Sodium periodate ≥99.8% ACS, Fluka™
Supplier: Honeywell Chemicals