Potassium Triphosphate, 94+% 1 * 250 g
Catalog # STRE93-1955-250G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Magnesium oxide light 98.0-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Citric acid, anhydrous ≥99.5%, powder, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium acetate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals