Sodium Hexachloroplatinate(Iv) Hexahydra 1 * 5 g
Catalog # STRE78-1995-5G
Supplier: Strem Chemicals, Inc.
Sodium Hexachloroplatinate(Iv) Hexahydra 1 * 5 g
Catalog # STRE78-1995-5G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Mercury(II) iodide, red Ph. Franç., extra pure, Supelco®
Supplier: Merck
Sodium Tellurate(Vi) Dihydrate, 99.5% 1 * 5 g
Supplier: Strem Chemicals, Inc.
Praseodymium(Iii) Phosphate 1 * 25 g
Supplier: Strem Chemicals, Inc.
Cesium Nitrate (99.99%-Cs) Puratrem 1 * 50 g
Supplier: Strem Chemicals, Inc.
Niobium Rod (99.8%) 1 * 25 cm
Supplier: Strem Chemicals, Inc.
Cobalt(Ii,Iii) Oxide, 99.5% 1 * 25 g
Supplier: Strem Chemicals, Inc.
AMMONIA AS NH3 - 1000 MG/L 1 * 125 mL
Supplier: WATERS
Iron(II) chloride tetrahydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Holmium Chips (99% Reo) 1 * 10 g
Supplier: Strem Chemicals, Inc.
Osmium(Viii) Oxide, 4% In Water 1 * 10 mL
Supplier: Strem Chemicals, Inc.
Rhenium(Vii) Oxide (99.99%-Re) Puratrem 1 * 1 g
Supplier: Strem Chemicals, Inc.
Antimony(Iii) Telluride (99.96%-Sb) 1 * 5 g
Supplier: Strem Chemicals, Inc.
Aluminium chloride, anhydrous 98.5%, powder, extra pure
Supplier: Thermo Fisher Scientific
Ammonium chloride ≥99.5% ACS
Supplier: VWR Chemicals
Disodium hydrogen phosphate dihydrate 99+% for HPLC
Supplier: Thermo Fisher Scientific
1-Pentanesulphonic acid sodium salt monohydrate 98+% for HPLC
Supplier: Thermo Fisher Scientific
Iron(Iii) Bromide, Anhydrous, 99% 1 * 10 g
Supplier: Strem Chemicals, Inc.